Merck
  • Chemical library and structure-activity relationships of 11-demethyl-12-oxo calanolide A analogues as anti-HIV-1 agents.
Chemical library and structure-activity relationships of 11-demethyl-12-oxo calanolide A analogues as anti-HIV-1 agents.
Journal of medicinal chemistry (2008-02-21)
Tao Ma, Li Liu, Hai Xue, Li Li, Chunyan Han, Lin Wang, Zhiwei Chen, Gang Liu
ZUSAMMENFASSUNG
(+)-Calanolide A ( 1) as a natural product was previously found as an inhibitor of HIV-1 reverse transcriptase. In our further investigation of its template, racemic 11-demethyl-12-oxo calanolide A ( 15), which had two fewer chiral carbon centers at the C-11 and C-12 positions than (+)-calanolide A, had a comparably inhibitory activity and better therapeutic index (EC 50 = 0.11 microM, TI = 818) against HIV-1 in vitro. A library based on its structural core was then designed and synthesized with introduction of nine diversity points in this article. The evaluations of anti-HIV-1 activity in vitro concluded their structure-activity relationships (SARs). A novel compound (10-bromomethyl-11-demethyl-12-oxo calanolide A, 123) was identified to have much higher inhibitory potency and therapeutic index (EC 50 = 2.85 nM, TI > 10,526) than those of the class compound against HIV-1. This finding provided a very important clue that modifications of the C ring at the C-10 position may be conducted to obtain drug candidates with better activity against HIV-1.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
1,1-Diethoxy-3-methyl-2-buten, 97%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.