Merck
  • Functional role of T-cell receptor nanoclusters in signal initiation and antigen discrimination.
Functional role of T-cell receptor nanoclusters in signal initiation and antigen discrimination.
Proceedings of the National Academy of Sciences of the United States of America (2016-08-31)
Sophie V Pageon, Thibault Tabarin, Yui Yamamoto, Yuanqing Ma, Philip R Nicovich, John S Bridgeman, André Cohnen, Carola Benzing, Yijun Gao, Michael D Crowther, Katie Tungatt, Garry Dolton, Andrew K Sewell, David A Price, Oreste Acuto, Robert G Parton, J Justin Gooding, Jérémie Rossy, Jamie Rossjohn, Katharina Gaus
ZUSAMMENFASSUNG
Antigen recognition by the T-cell receptor (TCR) is a hallmark of the adaptive immune system. When the TCR engages a peptide bound to the restricting major histocompatibility complex molecule (pMHC), it transmits a signal via the associated CD3 complex. How the extracellular antigen recognition event leads to intracellular phosphorylation remains unclear. Here, we used single-molecule localization microscopy to quantify the organization of TCR-CD3 complexes into nanoscale clusters and to distinguish between triggered and nontriggered TCR-CD3 complexes. We found that only TCR-CD3 complexes in dense clusters were phosphorylated and associated with downstream signaling proteins, demonstrating that the molecular density within clusters dictates signal initiation. Moreover, both pMHC dose and TCR-pMHC affinity determined the density of TCR-CD3 clusters, which scaled with overall phosphorylation levels. Thus, TCR-CD3 clustering translates antigen recognition by the TCR into signal initiation by the CD3 complex, and the formation of dense signaling-competent clusters is a process of antigen discrimination.