Merck
  • RGC-32 is expressed in the human atherosclerotic arterial wall: Role in C5b-9-induced cell proliferation and migration.
RGC-32 is expressed in the human atherosclerotic arterial wall: Role in C5b-9-induced cell proliferation and migration.
Experimental and molecular pathology (2016-09-14)
Sonia I Vlaicu, Alexandru Tatomir, Dallas Boodhoo, Takahiro Ito, Matthew Fosbrink, Cornelia Cudrici, Armugam P Mekala, Jonathan Ciriello, Doiniţa Crişan, Emil Boţan, Violeta Rus, Horea Rus
ZUSAMMENFASSUNG
The complement system is an important player in the development of atherosclerosis. Previously reported as a cell cycle regulator, RGC-32 is an essential effector of the terminal complement complex, C5b-9. In this study, our aims were to determine the expression of RGC-32 in the human atherosclerotic arterial wall and to delineate the mechanisms through which RGC-32 affects C5b-9-induced endothelial cell proliferation and migration. We now demonstrate that RGC-32 is expressed in human aortic atherosclerotic wall and that RGC-32 expression increases with the progression of atherosclerosis. Furthermore, silencing of RGC-32 expression abolished C5b-9-induced human aortic endothelial cell (HAEC) proliferation and migration. Of the 279 genes differentially expressed in HAECs after RGC-32 silencing, the genes involved in cell adhesion and cell cycle activation were significantly regulated by RGC-32. RGC-32 silencing caused a significant reduction in the expression of cyclin D1, cyclin D3, Akt, ROCK1, Rho GDP dissociation inhibitor alpha and profilin. These data suggest that RGC-32 mediates HAEC migration through the regulation of RhoA and ROCK1 expression and is involved in actin cytoskeletal organization. Thus, RGC-32 has promising therapeutic potential with regard to angiogenesis and atherosclerosis.