Merck
  • Attachment of an aminoglycoside, amikacin, to implantable collagen for local delivery in wounds.
Attachment of an aminoglycoside, amikacin, to implantable collagen for local delivery in wounds.
Antimicrobial agents and chemotherapy (1993-09-01)
S T Boyce, A P Supp, G D Warden, I A Holder
ZUSAMMENFASSUNG
Cultured skin substitutes consisting of implantable collagen (COL) and cultured human skin cells often fail clinically from destruction by microbial contamination. Hypothetically, addition of selected antimicrobial drugs to the implant may control microbial contamination and increase healing of skin wounds with these materials. As a model for drug delivery, bovine skin COL (1 mg/ml) and amikacin (AM; 46 micrograms/ml) were modified by covalent addition of biotin (B-COL and B-AM, respectively) from B-N-hydroxysuccinimide and bound together noncovalently with avidin (A). B-COL was incubated with A and then with B-peroxidase (B-P) or by serial incubation with B-AM and B-P, before P-dependent chromogen formation. Colorimetric data (n = 12 per condition) from spot tests on nitrocellulose paper were collected by transmission spectrophotometry. Specificity of drug binding in spot tests was determined by (i) serial dilution of B-COL; (ii) reactions with COL, AM, or P that had no B; (iii) removal of A; or (iv) preincubation of B-COL-A with B before incubation with B-P. Binding of B-AM was (i) dependent on the concentration of B-COL; (ii) specific to B-COL, A, and B-P (P < 0.05); and (iii) not eluted by incubation in 0.15 or 1.0 M NaCl. B-AM was found to block binding of B-P to the B-COL-A complex and to retain bacteriocidal activity against 10 clinical isolates of wound bacteria in the wet disc assay. Antimicrobial activity of B-AM was removed from solution by treatment with magnetic A and a permanent magnet. These results suggest that selected antimicrobial drugs can be biotinylated for attachments to COL-cultured cell implants without loss of pharmacologic activity. Because this chemistry utilizes a common ligand, any molar ratio of agents may be administered simultaneously and localized to the site of implantation.