Merck
  • Selective inhibition of IFNG-induced autophagy by Mir155- and Mir31-responsive WNT5A and SHH signaling.
Selective inhibition of IFNG-induced autophagy by Mir155- and Mir31-responsive WNT5A and SHH signaling.
Autophagy (2013-12-18)
Sahana Holla, Mariola Kurowska-Stolarska, Jagadeesh Bayry, Kithiganahalli Narayanaswamy Balaji
ZUSAMMENFASSUNG
Autophagy is one of the major immune mechanisms engaged to clear intracellular infectious agents. However, several pathogens have evolved strategies to evade autophagy. Here, we demonstrated that Mycobacteria, Shigella, and Listeria but not Klebsiella, Staphylococcus, and Escherichia inhibit IFNG-induced autophagy in macrophages by evoking selective and robust activation of WNT and SHH pathways via MTOR. Utilization of gain- or loss-of-function analyses as well as mir155-null macrophages emphasized the role of MTOR-responsive epigenetic modifications in the induction of Mir155 and Mir31. Importantly, cellular levels of PP2A, a phosphatase, were regulated by Mir155 and Mir31 to fine-tune autophagy. Diminished expression of PP2A led to inhibition of GSK3B, thus facilitating the prolonged activation of WNT and SHH signaling pathways. Sustained WNT and SHH signaling effectuated the expression of anti-inflammatory lipoxygenases, which in tandem inhibited IFNG-induced JAK-STAT signaling and contributed to evasion of autophagy. Altogether, these results established a role for new host factors and inhibitory mechanisms employed by the pathogens to limit autophagy, which could be targeted for therapeutic interventions.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Rapamycin, ≥95% (HPLC), powder
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Monoklonaler Anti-β-Aktin−Peroxidase-Antikörper der Maus in Maus hergestellte Antikörper, clone AC-15, purified from hybridoma cell culture
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
LY 294002, LY294002, CAS 154447-36-6, is a cell-permeable, potent, reversible, and specific inhibitor of PI 3-kinase ((IC50 = 1.4 µM). Acts on the ATP-binding site.
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Cyclopamin, V. californicum, Cyclopamine, V. californicum, CAS 4449-51-8, is a cell-permeable steroidal alkaloid & cholesterol mimic that specifically antagonizes Sonic Hedgehog signaling through direct interaction with Smo.
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Kalpain-Inhibitor I, ≥97% (TLC), powder
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Monoclonal Anti-AGO2 antibody produced in rat, ~1.5 mg/mL, clone 11A9, purified immunoglobulin
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Wnt-Antagonist II, IWP-2 – CAS 686770-61-6 – Calbiochem, The Wnt Antagonist II, IWP-2, also referenced under CAS 686770-61-6, controls the biological activity of Wnt. This small molecule/inhibitor is primarily used for Cancer applications.
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
ββ-Catenin/Tcf-Inhibitor, FH535, The β-Catenin/Tcf Inhibitor, FH535, also referenced under CAS 108409-83-2, controls the biological activity of β-Catenin/Tcf. This small molecule/inhibitor is primarily used for Cell Signaling applications.
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
AG 490, CAS-Nr. 133550-30-8, Calbiochem, A cell-permeable, reversible, substrate competitive, and potent inhibitor of epidermal growth factor receptor kinase autophosphorylation (IC50 = 100 nM).
Preise und Verfügbarkeit sind derzeit nicht verfügbar.