Merck
Physcion induces apoptosis in hepatocellular carcinoma by modulating miR-370.
American journal of cancer research (2017-01-04)
Xiaoping Pan, Haixia Wang, Dongmeng Tong, Chen Wang, Li Sun, Chunjuan Zhao, Yan Li, Lida Zhu, Di Wu
ZUSAMMENFASSUNG
Hepatocellular carcinoma (HCC) is one of the most common and aggressive malignancies. The current study is designed to explore the role of physcion, a major active ingredient in several traditional herbal medicinal plants, for the treatment of HCC. HCC cell lines, SMMC7721 and HepG2, were treated with physcion and its apoptosis-inducing effect was examined. Both in vitro and in vivo results from the present study demonstrated that physcion treatment resulted in apoptotic cell death in HCC cells via upregulation of miR-370. Furthermore, our findings showed that the physcion modulated the level of miR-370 through AMPK/Sp1/DNMT1 signaling. Taken together, these results showed that physcion exerts anti-tumor effect against HCC, which may be a potential agent for the adjunct chemotherapy.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
L-(−)-Glukose, ≥99%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
(Tyr[SO3H]27)Cholecystokinin-Fragment 26-33 Amid, ≥97% (HPLC), powder
Preise und Verfügbarkeit sind derzeit nicht verfügbar.