Merck
Catalytic enantioselective aldol reaction to ketones.
Journal of the American Chemical Society (2006-06-01)
Kounosuke Oisaki, Dongbo Zhao, Motomu Kanai, Masakatsu Shibasaki
ZUSAMMENFASSUNG
An enantioselective aldol reaction between ketones and ketene silyl acetals is described using CuF-chiral phosphine as a catalyst. The key for high enantioselectivity was the development of a novel ligand derived from Taniaphos combined with the unique accelerative effect of PhBF3K. These conditions are applicable to various substrates such as aromatic, aliphatic, and heteroaromatic ketones. In the case of substituted nucleophiles, the reaction proceeds well. The diastereoselectivity is independent of ketene silyl acetal geometry. This is the first example of a catalytic enantio- and diastereoselective aldol reaction to ketones using ketene silyl acetals.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Kupfer(II)-fluorid, 98%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.