Merck
  • Gas Phase Oxidation of Campholenic Aldehyde and Solution Phase Reactivity of its Epoxide Derivative.
Gas Phase Oxidation of Campholenic Aldehyde and Solution Phase Reactivity of its Epoxide Derivative.
The journal of physical chemistry. A (2016-12-13)
William C Thomas, William D Dresser, Diego A Cortés, Matthew J Elrod
ZUSAMMENFASSUNG
The rate constant for the OH reaction with campholenic aldehyde (CA) was measured using the flow tube-chemical ionization mass spectrometry method with a relative rate kinetics technique and was found to be (6.54 ± 0.52) × 10
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Iodmethan, contains copper as stabilizer, ReagentPlus®, 99%
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Glutaraldehyd -Lösung, Grade I, 70% in H2O, specially purified for use as an electron microscopy fixative or other sophisticated use
Preise und Verfügbarkeit sind derzeit nicht verfügbar.