Merck
  • Characterization of a new candidate isotopic reference material for natural Pb using primary measurement method.
Characterization of a new candidate isotopic reference material for natural Pb using primary measurement method.
Analytica chimica acta (2017-05-26)
Naoko Nonose, Toshihiro Suzuki, Ki-Cheol Shin, Tsutomu Miura, Akiharu Hioki
ZUSAMMENFASSUNG
A lead isotopic standard solution with natural abundance has been developed by applying a mixture of a solution of enriched
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Pyrvinium pamoate salt hydrate, ≥98% (HPLC)
Preise und Verfügbarkeit sind derzeit nicht verfügbar.