Merck
Exercise induces cerebral VEGF and angiogenesis via the lactate receptor HCAR1.
Nature communications (2017-05-24)
Cecilie Morland, Krister A Andersson, Øyvind P Haugen, Alena Hadzic, Liv Kleppa, Andreas Gille, Johanne E Rinholm, Vuk Palibrk, Elisabeth H Diget, Lauritz H Kennedy, Tomas Stølen, Eivind Hennestad, Olve Moldestad, Yiqing Cai, Maja Puchades, Stefan Offermanns, Koen Vervaeke, Magnar Bjørås, Ulrik Wisløff, Jon Storm-Mathisen, Linda H Bergersen
ZUSAMMENFASSUNG
Physical exercise can improve brain function and delay neurodegeneration; however, the initial signal from muscle to brain is unknown. Here we show that the lactate receptor (HCAR1) is highly enriched in pial fibroblast-like cells that line the vessels supplying blood to the brain, and in pericyte-like cells along intracerebral microvessels. Activation of HCAR1 enhances cerebral vascular endothelial growth factor A (VEGFA) and cerebral angiogenesis. High-intensity interval exercise (5 days weekly for 7 weeks), as well as L-lactate subcutaneous injection that leads to an increase in blood lactate levels similar to exercise, increases brain VEGFA protein and capillary density in wild-type mice, but not in knockout mice lacking HCAR1. In contrast, skeletal muscle shows no vascular HCAR1 expression and no HCAR1-dependent change in vascularization induced by exercise or lactate. Thus, we demonstrate that a substance released by exercising skeletal muscle induces supportive effects in brain through an identified receptor.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Natrium-L-lactat, ≥99.0% (NT)
Preise und Verfügbarkeit sind derzeit nicht verfügbar.