Merck
New opioid receptor antagonist: Naltrexone-14-O-sulfate synthesis and pharmacology.
European journal of pharmacology (2017-05-16)
Ferenc Zádor, Kornél Király, András Váradi, Mihály Balogh, Ágnes Fehér, Dóra Kocsis, Anna I Erdei, Erzsébet Lackó, Zoltán S Zádori, Sándor Hosztafi, Béla Noszál, Pál Riba, Sándor Benyhe, Susanna Fürst, Mahmoud Al-Khrasani
ZUSAMMENFASSUNG
Opioid antagonists, naloxone and naltrexone have long been used in clinical practice and research. In addition to their low selectivity, they easily pass through the blood-brain barrier. Quaternization of the amine group in these molecules, (e.g. methylnaltrexone) results in negligible CNS penetration. In addition, zwitterionic compounds have been reported to have limited CNS access. The current study, for the first time gives report on the synthesis and the in vitro [competition binding, G-protein activation, isolated mouse vas deferens (MVD) and mouse colon assay] pharmacology of the zwitterionic compound, naltrexone-14-O-sulfate. Naltrexone, naloxone, and its 14-O-sulfate analogue were used as reference compounds. In competition binding assays, naltrexone-14-O-sulfate showed lower affinity for µ, δ or κ opioid receptor than the parent molecule, naltrexone. However, the μ/κ opioid receptor selectivity ratio significantly improved, indicating better selectivity. Similar tendency was observed for naloxone-14-O-sulfate when compared to naloxone. Naltrexone-14-O-sulfate failed to activate [
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
[D-Ala2, N-Me-Phe4, Gly5-ol]-Enkephalin acetate salt, ≥97% (HPLC)
Sigma-Aldrich
[D-Pen2,5]-Enkephalin hydrate, ≥95% (HPLC)