Merck
  • RCAN1.4 regulates VEGFR-2 internalisation, cell polarity and migration in human microvascular endothelial cells.
RCAN1.4 regulates VEGFR-2 internalisation, cell polarity and migration in human microvascular endothelial cells.
Angiogenesis (2017-03-09)
Ahmad F Alghanem, Emma L Wilkinson, Maxine S Emmett, Mohammad A Aljasir, Katherine Holmes, Beverley A Rothermel, Victoria A Simms, Victoria L Heath, Michael J Cross
ZUSAMMENFASSUNG
Regulator of calcineurin 1 (RCAN1) is an endogenous inhibitor of the calcineurin pathway in cells. It is expressed as two isoforms in vertebrates: RCAN1.1 is constitutively expressed in most tissues, whereas transcription of RCAN1.4 is induced by several stimuli that activate the calcineurin-NFAT pathway. RCAN1.4 is highly upregulated in response to VEGF in human endothelial cells in contrast to RCAN1.1 and is essential for efficient endothelial cell migration and tubular morphogenesis. Here, we show that RCAN1.4 has a role in the regulation of agonist-stimulated VEGFR-2 internalisation and establishment of endothelial cell polarity. siRNA-mediated gene silencing revealed that RCAN1 plays a vital role in regulating VEGF-mediated cytoskeletal reorganisation and directed cell migration and sprouting angiogenesis. Adenoviral-mediated overexpression of RCAN1.4 resulted in increased endothelial cell migration. Antisense-mediated morpholino silencing of the zebrafish RCAN1.4 orthologue revealed a disrupted vascular development further confirming a role for the RCAN1.4 isoform in regulating vascular endothelial cell physiology. Our data suggest that RCAN1.4 plays a novel role in regulating endothelial cell migration by establishing endothelial cell polarity in response to VEGF.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Duolink® In Situ Detektionsreagenzien Rot
Sigma-Aldrich
Duolink® In Situ PLA® Sonde Anti-Kaninchen PLUS, Affinity purified Donkey anti-Rabbit IgG (H+L)
Sigma-Aldrich
Duolink® In Situ Rot Starterkit Maus/Kaninchen
Sigma-Aldrich
Duolink® In Situ PLA® Sonde Anti-Maus MINUS, Affinity purified Donkey anti-Mouse IgG (H+L)
Sigma-Aldrich
Duolink® In Situ PLA® Sonde Anti-Maus PLUS
Sigma-Aldrich
Duolink® In Situ PLA® Sonde Anti-Kaninchen MINUS
Sigma-Aldrich
Duolink® In-situ-Detektionsreagenzien Orange
Sigma-Aldrich
Duolink® In-situ-Detektionsreagenzien Tiefrot
Sigma-Aldrich
Duolink® In-situ-Detektionsreagenzien Grün
Sigma-Aldrich
Duolink® In Situ Waschpuffer, Fluoreszenz
Sigma-Aldrich
Natriumorthovanadat, ≥90% (titration)
Sigma-Aldrich
Duolink® In Situ Orange Starterkit Maus/Kaninchen
Sigma-Aldrich
Duolink® In Situ Eindeckmittel mit DAPI
Sigma-Aldrich
Aprotinin aus Rinderlunge, lyophilized powder, 3-7 TIU/mg solid
Sigma-Aldrich
Normale Kaninchen-IgG-Antikörper, Normal Rabbit IgG Polyclonal Antibody control validated for use in Immunoprecipitation & Western Blotting.
Sigma-Aldrich
Duolink® In Situ Probemaker PLUS
Sigma-Aldrich
Duolink® In Situ Probemaker MINUS
Sigma-Aldrich
Duolink® In Situ Detection Reagents Brightfield
Sigma-Aldrich
Duolink® flowPLA Detection Kit - FarRed, Duolink® PLA kit for Flow Cytometry with FarRed Detection
Sigma-Aldrich
Duolink® PLA Control Kit - PPI, Control Kit to reliably detect Protein-Protein Interactions (PPI) with Duolink® PLA
Sigma-Aldrich
Leupeptin, synthetic, ≥85% (HPLC)
Sigma-Aldrich
Duolink® In Situ PLA® Probe Anti-Goat MINUS, Affinity purified Donkey anti-Goat IgG (H+L)
Sigma-Aldrich
Duolink® In Situ Rot Starterkit Ziege/Kaninchen
Sigma-Aldrich
Duolink® In Situ Wash Buffer, Brightfield
Sigma-Aldrich
Duolink® In Situ PLA® Sonde Anti-Ziege PLUS
Sigma-Aldrich
Duolink® flowPLA Detection Kit - Red, DUOLINK® : PLA kit for Flow Cytometry with Red Detection
Sigma-Aldrich
Duolink®-flowPLA-Detektionskit – Grün, Duolink® PLA kit for Flow Cytometry with Green Detection
Sigma-Aldrich
Duolink® In Situ Red Starter Kit Mouse/Goat
Sigma-Aldrich
Duolink® In Situ Orange Starter Kit Goat/Rabbit
Sigma-Aldrich
Duolink® flowPLA Detection Kit - Orange, Duolink® PLA kit for Flow Cytometry with Orange Detection