Merck
  • Characterization of a novel theme C glycoside hydrolase family 9 cellulase and its CBM-chimeric enzymes.
Characterization of a novel theme C glycoside hydrolase family 9 cellulase and its CBM-chimeric enzymes.
Applied microbiology and biotechnology (2017-05-30)
Cheng-Jie Duan, Ming-Yue Huang, Hao Pang, Jing Zhao, Chao-Xing Wu, Jia-Xun Feng
ZUSAMMENFASSUNG
In bacterial cellulase systems, glycoside hydrolase family 9 (GH9) cellulases are generally regarded as the major cellulose-degrading factors besides GH48 exoglucanase. In this study, umcel9A, which was cloned from uncultured microorganisms from compost, with the encoded protein being theme C GH9 cellulase, was heterologously expressed in Escherichia coli, and the biochemical properties of the purified enzyme were characterized. Hydrolysis of carboxylmethylcellulose (CMC) by Umcel9A led to the decreased viscosity of CMC solution and production of reducing sugars. Interestingly, cellobiose was the major product when cellulosic materials were hydrolyzed by Umcel9A. Six representative carbohydrate-binding modules (CBMs) from different CBM families (CBM1, CBM2, CBM3, CBM4, CBM10, and CBM72) were fused with Umcel9A at the natural terminal position, resulting in significant enhancement of the binding capacity of the chimeric enzymes toward four different insoluble celluloses as compared with that of Umcel9A. Catalytic activity of the chimeric enzymes against insoluble celluloses, including phosphoric acid-swollen cellulose (PASC), alkali-pretreated sugarcane bagasse (ASB), filter paper powder (FPP), and Avicel, was higher than that of Umcel9A, except for Umcel9A-CBM3. In these chimeric enzymes, CBM4-Umcel9A exhibited the highest activity toward the four tested insoluble celluloses and displayed 4.2-, 3.0-, 2.4-, and 6.6-fold enhanced activity toward PASC, ASB, FPP, and Avicel, respectively, when compared with that of Umcel9A. CBM4-Umcel9A also showed highest V
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Cellotetraose, ≥85% (HPLC)
Preise und Verfügbarkeit sind derzeit nicht verfügbar.