Merck
Glioblastoma Therapy Can Be Augmented by Targeting IDH1-Mediated NADPH Biosynthesis.
Cancer research (2016-12-08)
Daniel R Wahl, Joseph Dresser, Kari Wilder-Romans, Joshua D Parsels, Shuang G Zhao, Mary Davis, Lili Zhao, Maureen Kachman, Stefanie Wernisch, Charles F Burant, Meredith A Morgan, Felix Y Feng, Corey Speers, Costas A Lyssiotis, Theodore S Lawrence
ZUSAMMENFASSUNG
NADPH is a critical reductant needed in cancer cells to fuel the biosynthesis of deoxynucleotides and antioxidants and to sustain stress-survival responses after radiation-induced DNA damage. Thus, one rational strategy to attack cancer cells is to target their heavy reliance on NADPH. Here, we report that the isocitrate dehydrogenase IDH1 is the most strongly upregulated NADPH-producing enzyme in glioblastoma (GBM). IDH1 silencing in GBM cells reduced levels of NADPH, deoxynucleotides, and glutathione and increased their sensitivity to radiation-induced senescence. Rescuing these metabolic restrictions was sufficient to reverse IDH1-mediated radiosensitization. In a murine xenograft model of human GBM, we found that IDH1 silencing significantly improved therapeutic responses to fractionated radiotherapy, when compared with either treatment alone. In summary, our work offers a mechanistic rationale for IDH1 inhibition as a metabolic strategy to improve the response of GBM to radiotherapy.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
MISSION® esiRNA, targeting human IDH1
Preise und Verfügbarkeit sind derzeit nicht verfügbar.