- Bioethanol Production from
Bioethanol Production from
Journal of microbiology and biotechnology (2017-04-04)
Seul Ki Kim, Cuong Mai Nguyen, Eun Hye Ko, In-Chul Kim, Jin-Seog Kim, Jin-Cheol Kim
PMID28372036
ZUSAMMENFASSUNG
The aim of this study was to develop a potential process for bioethanol production from