Merck
Ribosomal transcription is regulated by PGC-1alpha and disturbed in Huntington's disease.
Scientific reports (2017-08-19)
Sarah Jesse, Hanna Bayer, Marius C Alupei, Martina Zügel, Medhanie Mulaw, Francesca Tuorto, Silke Malmsheimer, Karmveer Singh, Jürgen Steinacker, Uwe Schumann, Albert C Ludolph, Karin Scharffetter-Kochanek, Anke Witting, Patrick Weydt, Sebastian Iben
ZUSAMMENFASSUNG
PGC-1α is a versatile inducer of mitochondrial biogenesis and responsive to the changing energy demands of the cell. As mitochondrial ATP production requires proteins that derive from translation products of cytosolic ribosomes, we asked whether PGC-1α directly takes part in ribosomal biogenesis. Here, we show that a fraction of cellular PGC-1α localizes to the nucleolus, the site of ribosomal transcription by RNA polymerase I. Upon activation PGC-1α associates with the ribosomal DNA and boosts recruitment of RNA polymerase I and UBF to the rDNA promoter. This induces RNA polymerase I transcription under different stress conditions in cell culture and mouse models as well as in healthy humans and is impaired already in early stages of human Huntington's disease. This novel molecular link between ribosomal and mitochondrial biogenesis helps to explain sarcopenia and cachexia in diseases of neurodegenerative origin.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Millipore
ANTI-FLAG® M2-Affinitätsgel, purified immunoglobulin, buffered aqueous glycerol solution
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Nα-Acetyl-L-lysin
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Anti-PGC-1-Antikörper, Chemicon®, from rabbit
Preise und Verfügbarkeit sind derzeit nicht verfügbar.