Merck
  • Acetylation of hMOF Modulates H4K16ac to Regulate DNA Repair Genes in Response to Oxidative Stress.
Acetylation of hMOF Modulates H4K16ac to Regulate DNA Repair Genes in Response to Oxidative Stress.
International journal of biological sciences (2017-08-16)
Jianing Zhong, Liying Ji, Huiqian Chen, Xianfeng Li, Jian'an Zhang, Xingxing Wang, Weilin Wu, Ying Xu, Fei Huang, Wanshi Cai, Zhong Sheng Sun
ZUSAMMENFASSUNG
Oxidative stress is considered to be a key risk state for a variety of human diseases. In response to oxidative stress, the regulation of transcriptional expression of DNA repair genes would be important to DNA repair and genomic stability. However, the overall pattern of transcriptional expression of DNA repair genes and the underlying molecular response mechanism to oxidative stress remain unclear. Here, by employing colorectal cancer cell lines following exposure to hydrogen peroxide, we generated expression profiles of DNA repair genes via RNA-seq and identified gene subsets that are induced or repressed following oxidative stress exposure. RRBS-seq analyses further indicated that transcriptional regulation of most of the DNA repair genes that were induced or repressed is independent of their DNA methylation status. Our analyses also indicate that hydrogen peroxide induces deacetylase SIRT1 which decreases chromatin affinity and the activity of histone acetyltransferase hMOF toward H4K16ac and results in decreased transcriptional expression of DNA repair genes. Taken together, our findings provide a potential mechanism by which oxidative stress suppresses DNA repair genes which is independent of the DNA methylation status of their promoters.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Anti-β-Actin-Antikörper, Maus monoklonal, clone AC-15, purified from hybridoma cell culture
Sigma-Aldrich
Anti-Maus-IgG (Gesamtmolekül)-Peroxidase in Ziege hergestellte Antikörper, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Trichostatin A, ≥98% (HPLC), from Streptomyces sp.
Sigma-Aldrich
Bleomycinsulfat aus Streptomyces verticillus, crystalline, 1.5-2.0 U/mg
Sigma-Aldrich
Anti-Kaninchen-IgG-(Gesamtmolekül)-Peroxidase in Ziege hergestellte Antikörper, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
(S)-(+)-Camptothecin, ≥90% (HPLC), powder
Sigma-Aldrich
Anti-Phospho-Histon H2A.X (Ser139)-Antikörper, Klon JBW301, Biotin-Konjugat, clone JBW301, Upstate®, from mouse
Sigma-Aldrich
Wortmannin aus Penicillium funiculosum, from Penicillium funiculosum, ≥98% (HPLC and TLC)
Sigma-Aldrich
Anti-acetyl-Histon H4 (Lys16)-Antikörper, Upstate®, from rabbit
Sigma-Aldrich
Anti-Trimethyl-Histon-H3(-Lys4)-Antikörper, Klon MC315, monoklonaler Kaninchen-Antikörper, culture supernatant, clone MC315, Upstate®
Sigma-Aldrich
Anti-Dimethyl-Histon-H3-(Lys4-)Antikörper, Klon CMA303, clone CMA303, from mouse
Sigma-Aldrich
NU7026, ≥98% (HPLC), solid
Sigma-Aldrich
Anti-Sirt1 Antibody, clone 3H10.2, clone 3H10.2, Upstate®, from mouse
Sigma-Aldrich
CGK733, ≥98% (HPLC), solid