Merck
  • Myoendothelial gap junctions mediate regulation of angiopoietin-2-induced vascular hyporeactivity after hypoxia through connexin 43-gated cAMP transfer.
Myoendothelial gap junctions mediate regulation of angiopoietin-2-induced vascular hyporeactivity after hypoxia through connexin 43-gated cAMP transfer.
American journal of physiology. Cell physiology (2017-06-24)
Jing Xu, Guangming Yang, Tao Li, Liangming Liu
ZUSAMMENFASSUNG
Angiopoietin-2 (Ang-2) contributes to vascular hyporeactivity after hemorrhagic shock and hypoxia through upregulation of inducible nitric oxide synthase (iNOS) in a vascular endothelial cell (VEC)-specific and Ang-2/Tie2 receptor-dependent manner. While iNOS is primarily expressed in vascular smooth muscle cells (VSMCs), the mechanisms of signal transfer from VECs to VSMCs are unknown. A double-sided coculture model with VECs and VSMCs from Sprague-Dawley rats was used to investigate the role of myoendothelial gap junctions (MEGJs), the connexin (Cx) isoforms involved, and other relevant mechanisms. After hypoxia, VSMCs treated with exogenous Ang-2 showed increased iNOS expression and hyporeactivity, as well as MEGJ formation and communication. These Ang-2 effects were suppressed by the MEGJ inhibitor 18α-glycyrrhetic acid (18-GA),
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
MISSION® esiRNA, targeting human ANGPT2
Preise und Verfügbarkeit sind derzeit nicht verfügbar.