- Hydrogen-rich medium protects mouse embryonic fibroblasts from oxidative stress by activating LKB1-AMPK-FoxO1 signal pathway.
Hydrogen-rich medium protects mouse embryonic fibroblasts from oxidative stress by activating LKB1-AMPK-FoxO1 signal pathway.
Biochemical and biophysical research communications (2017-07-27)
Jihyun Lee, Goowon Yang, Young-Joo Kim, Quynh Hoa Tran, Wonchae Choe, Insug Kang, Sung Soo Kim, Joohun Ha
PMID28743498
ZUSAMMENFASSUNG
Persistent oxidative stress is recognized as a major cause of many pathological conditions as well as ageing. However, most clinical trials of dietary antioxidants have failed to produce successful outcomes in treating oxidative stress-induced diseases. Molecular hydrogen (H
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Preise und Verfügbarkeit sind derzeit nicht verfügbar.