Merck
  • The Arabidopsis MOS4-Associated Complex Promotes MicroRNA Biogenesis and Precursor Messenger RNA Splicing.
The Arabidopsis MOS4-Associated Complex Promotes MicroRNA Biogenesis and Precursor Messenger RNA Splicing.
The Plant cell (2017-09-28)
Tianran Jia, Bailong Zhang, Chenjiang You, Yong Zhang, Liping Zeng, Shengjun Li, Kaeli C M Johnson, Bin Yu, Xin Li, Xuemei Chen
ZUSAMMENFASSUNG
In
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Monoklonaler Anti-α-Tubulin-Antikörper in Maus hergestellte Antikörper, clone DM1A, ascites fluid
Roche
Anti-GFP, from mouse IgG1κ (clones 7.1 and 13.1)