Merck
  • Split GFP technologies to structurally characterize and quantify functional biomolecular interactions of FTD-related proteins.
Split GFP technologies to structurally characterize and quantify functional biomolecular interactions of FTD-related proteins.
Scientific reports (2017-10-27)
Chiara Foglieni, Stéphanie Papin, Agnese Salvadè, Tariq Afroz, Sandra Pinton, Giona Pedrioli, Giorgio Ulrich, Magdalini Polymenidou, Paolo Paganetti
ZUSAMMENFASSUNG
Protein multimerization in physiological and pathological conditions constitutes an intrinsic trait of proteins related to neurodegeneration. Recent evidence shows that TDP-43, a RNA-binding protein associated with frontotemporal dementia and amyotrophic lateral sclerosis, exists in a physiological and functional nuclear oligomeric form, whose destabilization may represent a prerequisite for misfolding, toxicity and subsequent pathological deposition. Here we show the parallel implementation of two split GFP technologies, the GFP bimolecular and trimolecular fluorescence complementation (biFC and triFC) in the context of TDP-43 self-assembly. These techniques coupled to a variety of assays based on orthogonal readouts allowed us to define the structural determinants of TDP-43 oligomerization in a qualitative and quantitative manner. We highlight the versatility of the GFP biFC and triFC technologies for studying the localization and mechanisms of protein multimerization in the context of neurodegeneration.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
3,3′,5,5′-3,3,5,5-Tetramethylbenzidin (TMB) Flüssigsubstratsystem für ELISA, peroxidase substrate
Sigma-Aldrich
RIPA-Puffer
Sigma-Aldrich
Poly-D-Lysin -hydrobromid, mol wt 70,000-150,000, lyophilized powder, γ-irradiated, BioReagent, suitable for cell culture
Sigma-Aldrich
Albumin aus Rinderserum, cold ethanol fraction, pH 5.2, ≥96%
Sigma-Aldrich
SIGMAFAST Proteaseinhibitor-Tabletten, For General Use
C17.2, NOTE: Both the cell line and DNA from the cell line may be available for this product. Please choose -1VL or VIAL for cells, or -DNA-5UG for DNA, 7062902, mouse brain, Neuronal
Zelllinie SH-SY5Y human, NOTE: Both the cell line and DNA from the cell line may be available for this product. Please choose -1VL or VIAL for cells, or -DNA-5UG for DNA, 94030304, human nerve, Neuroblast