- Binding and Signaling Studies Disclose a Potential Allosteric Site for Cannabidiol in Cannabinoid CB
Binding and Signaling Studies Disclose a Potential Allosteric Site for Cannabidiol in Cannabinoid CB
Frontiers in pharmacology (2017-11-08)
Eva Martínez-Pinilla, Katia Varani, Irene Reyes-Resina, Edgar Angelats, Fabrizio Vincenzi, Carlos Ferreiro-Vera, Julen Oyarzabal, Enric I Canela, José L Lanciego, Xavier Nadal, Gemma Navarro, Pier Andrea Borea, Rafael Franco
PMID29109685
ZUSAMMENFASSUNG
The mechanism of action of cannabidiol (CBD), the main non-psychotropic component of
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Preise und Verfügbarkeit sind derzeit nicht verfügbar.