- Effect of pressure on bilayer phase behavior of N-methylated di-O-hexadecylphosphatidylethanolamines: relevance of head-group modification on the bilayer interdigitation.
Effect of pressure on bilayer phase behavior of N-methylated di-O-hexadecylphosphatidylethanolamines: relevance of head-group modification on the bilayer interdigitation.
Biophysical chemistry (2017-04-16)
Masaki Goto, Yuya Aoki, Nobutake Tamai, Hitoshi Matsuki
PMID28410942
ZUSAMMENFASSUNG
The phase transitions of N-methylated di-O-hexadecylphosphatidylethanolamines (DHPE, DH-N-methyl-PE (DHMePE) and DH-N,N-dimethyl-PE (DHMe
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
N,N-Dimethyl-6-propionyl-2-naphthylamin, BioReagent, suitable for fluorescence, ≥98.0% (HPLC)
Preise und Verfügbarkeit sind derzeit nicht verfügbar.