Merck
  • CENP-B protects centromere chromatin integrity by facilitating histone deposition via the H3.3-specific chaperone Daxx.
CENP-B protects centromere chromatin integrity by facilitating histone deposition via the H3.3-specific chaperone Daxx.
Epigenetics & chromatin (2017-12-24)
Viacheslav M Morozov, Serena Giovinazzi, Alexander M Ishov
ZUSAMMENFASSUNG
The main chromatin unit, the nucleosome, can be modulated by the incorporation of histone variants that, in combination with posttranslational histones modifications, determine epigenetics properties of chromatin. Understanding the mechanism that creates a histone variants landscape at different genomic elements is expected to elevate our comprehension of chromatin assembly and function. The Daxx chaperone deposits transcription-associated histone H3.3 at centromeres, but mechanism of centromere-specific Daxx targeting remains unclear. In this study, we identified an unexpected function of the constitutive centromeric protein CENP-B that serves as a "beacon" for H3.3 incorporation. CENP-B depletion reduces Daxx association and H3.3 incorporation at centromeres. Daxx/CENP-B interaction and Daxx centromeric association are SUMO dependent and requires SIMs of Daxx. Depletion of SUMO-2, but not SUMO-1, decreases Daxx/CENP-B interaction and reduces centromeric accumulation of Daxx and H3.3, demonstrating distinct functions of SUMO paralogs in H3.3 chaperoning. Finally, disruption of CENP-B/Daxx-dependent H3.3 pathway deregulates heterochromatin marks H3K9me3, ATRX and HP1α at centromeres and elevates chromosome instability. The demonstrated roles of CENP-B and SUMO-2 in H3.3 loading reveal a novel mechanism controlling chromatin maintenance and genome stability. Given that CENP-B is the only centromere protein that binds centromere-specific DNA elements, our study provides a new link between centromere DNA and unique epigenetic landscape of centromere chromatin.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Leupeptin, microbial, ≥90% (HPLC)
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Pepstatin A, microbial, ≥90% (HPLC)
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Z-Leu-Leu-Leu-al, ≥90% (HPLC)
Preise und Verfügbarkeit sind derzeit nicht verfügbar.