- Maghemite Nanoparticles Acts as Nanozymes, Improving Growth and Abiotic Stress Tolerance in Brassica napus.
Maghemite Nanoparticles Acts as Nanozymes, Improving Growth and Abiotic Stress Tolerance in Brassica napus.
Nanoscale research letters (2017-12-21)
N G Martin Palmqvist, Gulaim A Seisenbaeva, Peter Svedlindh, Vadim G Kessler
PMID29260423
ZUSAMMENFASSUNG
Yttrium doping-stabilized γ-Fe