Merck
  • Glyoxal as an alternative fixative to formaldehyde in immunostaining and super-resolution microscopy.
Glyoxal as an alternative fixative to formaldehyde in immunostaining and super-resolution microscopy.
The EMBO journal (2017-11-18)
Katharina N Richter, Natalia H Revelo, Katharina J Seitz, Martin S Helm, Deblina Sarkar, Rebecca S Saleeb, Elisa D'Este, Jessica Eberle, Eva Wagner, Christian Vogl, Diana F Lazaro, Frank Richter, Javier Coy-Vergara, Giovanna Coceano, Edward S Boyden, Rory R Duncan, Stefan W Hell, Marcel A Lauterbach, Stephan E Lehnart, Tobias Moser, Tiago F Outeiro, Peter Rehling, Blanche Schwappach, Ilaria Testa, Bolek Zapiec, Silvio O Rizzoli
ZUSAMMENFASSUNG
Paraformaldehyde (PFA) is the most commonly used fixative for immunostaining of cells, but has been associated with various problems, ranging from loss of antigenicity to changes in morphology during fixation. We show here that the small dialdehyde glyoxal can successfully replace PFA Despite being less toxic than PFA, and, as most aldehydes, likely usable as a fixative, glyoxal has not yet been systematically tried in modern fluorescence microscopy. Here, we tested and optimized glyoxal fixation and surprisingly found it to be more efficient than PFA-based protocols. Glyoxal acted faster than PFA, cross-linked proteins more effectively, and improved the preservation of cellular morphology. We validated glyoxal fixation in multiple laboratories against different PFA-based protocols and confirmed that it enabled better immunostainings for a majority of the targets. Our data therefore support that glyoxal can be a valuable alternative to PFA for immunostaining.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Anti-β-Actin-Antikörper, Maus monoklonal, clone AC-15, purified from hybridoma cell culture
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Paraformaldehyd, reagent grade, crystalline
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Anti-Maus-IgG - Atto 647N, contains 50% glycerol as stabilizer
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Monoclonal Anti-TOMM20 antibody produced in mouse, clone 4F3, purified immunoglobulin, buffered aqueous solution
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
α-Bungarotoxin-tetramethylrhodamine from Bungarus multicinctus (Formosan Banded Krait)
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
1,2-Dilinoleoyl-3-palmitoyl-rac-Glycerin, ≥95% (TLC), liquid
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
SynaptoGreen C4, ≥95% (HPLC), solid
Preise und Verfügbarkeit sind derzeit nicht verfügbar.