Merck
  • The Ribonucleic Complex HuR-MALAT1 Represses CD133 Expression and Suppresses Epithelial-Mesenchymal Transition in Breast Cancer.
The Ribonucleic Complex HuR-MALAT1 Represses CD133 Expression and Suppresses Epithelial-Mesenchymal Transition in Breast Cancer.
Cancer research (2016-05-20)
Elisa Latorre, Stephana Carelli, Ivan Raimondi, Vito D'Agostino, Ilaria Castiglioni, Chiara Zucal, Giacomina Moro, Andrea Luciani, Giorgio Ghilardi, Eleonora Monti, Alberto Inga, Anna Maria Di Giulio, Alfredo Gorio, Alessandro Provenzani
ZUSAMMENFASSUNG
Epithelial-to-mesenchymal transition (EMT) is a core process underlying cell movement during embryonic development and morphogenesis. Cancer cells hijack this developmental program to execute a multi-step cascade, leading to tumorigenesis and metastasis. CD133 (PROM1), a marker of cancer stem cells, has been shown to facilitate EMT in various cancers, but the regulatory networks controlling CD133 gene expression and function in cancer remain incompletely delineated. In this study, we show that a ribonucleoprotein complex including the long noncoding RNA MALAT1 and the RNA-binding protein HuR (ELAVL1) binds the CD133 promoter region to regulate its expression. In luminal nonmetastatic MCF-7 breast cancer cells, HuR silencing was sufficient to upregulate N-cadherin (CDH2) and CD133 along with a migratory and mesenchymal-like phenotype. Furthermore, we found that in the basal-like metastatic cell line MDA-MB-231 and primary triple-negative breast cancer tumor cells, the repressor complex was absent from the CD133-regulatory region, but was present in the MCF-7 and primary ER+ tumor cells. The absence of the complex from basal-like cells was attributed to diminished expression of MALAT1, which, when overexpressed, dampened CD133 levels. In conclusion, our findings suggest that the failure of a repressive complex to form or stabilize in breast cancer promotes CD133 upregulation and an EMT-like program, providing new mechanistic insights underlying the control of prometastatic processes. Cancer Res; 76(9); 2626-36. ©2016 AACR.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Monoklonales Anti-β-Aktin in Maus hergestellte Antikörper, clone AC-15, ascites fluid
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Cyclosporin A, 97.0-101.5% (on dried basis)
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
Anti-Zellkern-Antikörper, Klon 235-1, clone 235-1, Chemicon®, from mouse
Preise und Verfügbarkeit sind derzeit nicht verfügbar.
Sigma-Aldrich
QCM Chemotaxis-Zellmigrationsassay, 24-Well (8 µm), fluorimetrisch, The QCM 24-well Migration Assay is ideal for the study of chemotaxis cell migration. The assay uses a 24-well plate with an 8 micron pore size, with fluorescent detection.
Preise und Verfügbarkeit sind derzeit nicht verfügbar.