Merck
  • Oroxylin A prevents angiogenesis of LSECs in liver fibrosis via inhibition of YAP/HIF-1α signaling.
Oroxylin A prevents angiogenesis of LSECs in liver fibrosis via inhibition of YAP/HIF-1α signaling.
Journal of cellular biochemistry (2017-09-01)
Chenxi Zhang, Mianli Bian, Xingran Chen, Huanhuan Jin, Shifeng Zhao, Xiang Yang, Jiangjuan Shao, Anping Chen, Qinglong Guo, Feng Zhang, Shizhong Zheng
ZUSAMMENFASSUNG
Angiogenesis of liver sinusoidal endothelial cells (LSECs) accompanies with hypoxia in liver fibrosis and they are of mutual promotion, which has raised wide concern. Here we established murine model of liver fibrosis and found that oroxylin A (40 mg/kg) could ameliorate angiogenesis in liver fibrosis may related to hypoxia inducible factor 1α (HIF-1α). The underlying mechanism was further investigated by isolating and culturing murine primary LSECs. Hypoxia induced vascular endothelial growth factor A (VEGF-A), angiopoietin 2 (Ang-2), and platelet endothelial cell adhesion molecule-1 (PECAM-1/CD31) elevated in LSECs were reduced by oroxylin A or acriflavine (ACF, an HIF-1α inhibitor), indicating HIF-1α involved the angiogenesis of LSECs. Additionally, interference with Yes-associated protein (YAP) significant downregulated the protein expression of HIF-1α and VEGF-A, while YAP plasmid exhibited an opposite effect. We next found that oroxylin A inhibited hypoxia-induced nuclear translocation of YAP, which may influence the accumulation of HIF-1α and subsequently decrease transcription of downstream target gene including VEGF-A and Ang-2, thereby exerting an anti-angiogenic activity.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Edelfosine, ≥95% (HPLC)
Preise und Verfügbarkeit sind derzeit nicht verfügbar.