Merck
  • Modifying culture conditions in chemical library screening identifies alternative inhibitors of mycobacteria.
Modifying culture conditions in chemical library screening identifies alternative inhibitors of mycobacteria.
Antimicrobial agents and chemotherapy (2009-09-30)
Christopher H Miller, Shahista Nisa, Sandi Dempsey, Cameron Jack, Ronan O'Toole
ZUSAMMENFASSUNG
In this study, application of a dual absorbance/fluorescence assay to a chemical library screen identified several previously unknown inhibitors of mycobacteria. In addition, growth conditions had a significant effect on the activity profile of the library. Some inhibitors such as Se-methylselenocysteine were detected only when screening was performed under nutrient-limited culture conditions as opposed to nutrient-rich culture conditions. We propose that multiple culture condition library screening is required for complete inhibitory profiling and for maximal antimycobacterial compound detection.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Kalzium-Ionophor A23187, ≥98% (TLC), powder
Sigma-Aldrich
Rifampicin, ≥95% (HPLC), powder or crystals
Sigma-Aldrich
1,10-Phenanthrolin, ≥99%
Sigma-Aldrich
Ciprofloxacin, ≥98% (HPLC)
Sigma-Aldrich
Tetracyclin, 98.0-102.0% (HPLC)
Sigma-Aldrich
Ofloxacin, fluoroquinolone antibiotic
Sigma-Aldrich
Rifampicin, suitable for plant cell culture, BioReagent, ≥95% (HPLC), powder or crystals
Sigma-Aldrich
Clotrimazol
Sigma-Aldrich
LOPAC®1280 – Kleiner Maßstab, International Version
Supelco
Ciprofloxacin, VETRANAL®, analytical standard
Sigma-Aldrich
Calmidazoliumchlorid, solid
Supelco
Ofloxacin, VETRANAL®, analytical standard
Sigma-Aldrich
LOPAC®1280 (International Version)