MET-Biodiesel Capillary GC Column
MET-Biodiesel Capillary GC Column
The MET-Biodiesel was designed specifically for the determination of free and total glycerin in B100 biodiesel samples.
Features and benefits of this column include:
- Good peak shape and resolution for all glyceride impurities of interest
- Ability to separate glycerin in addition to mono-, di-, and triglycerides (as methyl esters) from the FAMEs
- A maximum temperature of 380 °C (isothermal) and 430 °C (programmed), exceeding the temperature limitations specified in biodiesel methods ASTM® D6584 and EN 14105:2003
- Metal was selected as the column material because it holds up better than fused silica under the method conditions, virtually eliminating column breakage
- Integrated guard protects the analytical column from excess reagent and non-volatile compounds, extending column life with a leak-free connection
Chromatograms
The following chromatograms were generated using the MET-Biodiesel: (click to view)
| Description | Cat. No. |
|---|---|
| MET-Biodiesel, 14 m x 0.53 mm I.D., 0.16 µm with integrated 2 m x 0.53 mm I.D. guard | 28668-U |
Blank Containing Internal Standards Only on the MET-Biodiesel
| Peak IDs | |
|---|---|
| 1. | Butanetriol (I.S.) |
| 2. | Tricaprin (I.S.) |
| Conditions | |
|---|---|
| Column: | MET-Biodiesel, 14 m x 0.53 mm I.D., 0.16 µM with integrated 2 m x 0.53 mm I.D. guard (28668-U) |
| Oven: | 50 °C (1 min.), 15 °C/min. to 180 °C, 7 °C/min. to 230 °C, 30 °C/min. to 380 °C (10 min.) |
| Det.: | FID, 380 °C |
| Carrier Gas: | helium, 3.0 mL/min. |
| Injection: | 1 µL, cold on-column |
| Sample: | Method blank consisting of Butanetriol Internal Standard (44896-U) and Tricaprin Internal Standard (44897-U), derivatized with MSTFA (394866) then diluted in heptane |
ASTM® D6584 Calibration Standard on the MET-Biodiesel, Run 1 on a New Column
| Peak IDs | |
|---|---|
| 1. | Glycerin |
| 2. | Butanetriol (I.S.) |
| 3. | Monoolein |
| 4. | Tricaprin (I.S.) |
| 5. | Diolein |
| 6. | Triolein |
| Conditions | |
|---|---|
| Column: | MET-Biodiesel, 14 m x 0.53 mm I.D., 0.16 µM with integrated 2 m x 0.53 mm I.D. guard (28668-U) |
| Oven: | 50 °C (1 min.), 15 °C/min. to 180 °C, 7 °C/min. to 230 °C, 30 °C/min. to 380 °C (10 min.) |
| Det.: | FID, 380 °C |
| Carrier Gas: | helium, 3.0 mL/min. |
| Injection: | 1 µL, cold on-column |
| Sample: | ASTM® D6584 Standard Solution 1 (44899-U) plus Butanetriol Internal Standard (44896-U) and Tricaprin Internal Standard (44897-U), derivatized with MSTFA (394866) then diluted in heptane |
ASTM® D6584 Calibration Standard on the MET-Biodiesel, Run 100 on the Same Column
| Peak IDs | |
|---|---|
| 1. | Glycerin |
| 2. | Butanetriol (I.S.) |
| 3. | Monoolein |
| 4. | Tricaprin (I.S.) |
| 5. | Diolein |
| 6. | Triolein |
| Conditions | |
|---|---|
| Column: | MET-Biodiesel, 14 m x 0.53 mm I.D., 0.16 µM with integrated 2 m x 0.53 mm I.D. guard (28668-U) |
| Oven: | 50 °C (1 min.), 15 °C/min. to 180 °C, 7 °C/min. to 230 °C, 30 °C/min. to 380 °C (10 min.) |
| Det.: | FID, 380 °C |
| Carrier Gas: | helium, 3.0 mL/min. |
| Injection: | 1 µL, cold on-column |
| Sample: | ASTM® D6584 Standard Solution 1 (44899-U) plus Butanetriol Internal Standard (44896-U) and Tricaprin Internal Standard (44897-U), derivatized with MSTFA (394866) then diluted in heptane |
EN 14105 Monoglyceride Standard on the MET-Biodiesel
| Peak IDs | |
|---|---|
| 1. | Butanetriol (I.S.) |
| 2. | Monopalmitin (C16:0) |
| 3. | Monoolein (C18:1) |
| 4. | Monostearin (C18:0) |
| 5. | Tricaprin (I.S.) |
| Conditions | |
|---|---|
| Column: | MET-Biodiesel, 14 m x 0.53 mm I.D., 0.16 µM with integrated 2 m x 0.53 mm I.D. guard (28668-U) |
| Oven: | 50 °C (1 min.), 15 °C/min. to 180 °C, 7 °C/min. to 230 °C, 30 °C/min. to 380 °C (10 min.) |
| Det.: | FID, 380 °C |
| Carrier Gas: | helium, 3.0 mL/min. |
| Injection: | 1 µL, cold on-column |
| Sample: | EN 14105:2003 Monoglyceride Stock Solution (49446-U) plus Butanetriol Internal Standard (44896-U) and Tricaprin Internal Standard (44897-U), derivatized with MSTFA (394866) then diluted in heptane |
B100 Biodiesel Sample on the MET-Biodiesel
| Peak IDs | |
|---|---|
| 1. | Glycerin |
| 2. | Butanetriol (I.S.) |
| 3. | Monopalmitin (C16:0) |
| 4. | Monoolein (C18:1), Monolinolein (C18:2), Monolinolenin (C18:3) |
| 5. | Monostearin (C18:0) |
| 6. | Tricaprin (I.S.) |
| Conditions | |
|---|---|
| Column: | MET-Biodiesel, 14 m x 0.53 mm I.D., 0.16 µM with integrated 2 m x 0.53 mm I.D. guard (28668-U) |
| Oven: | 50 °C (1 min.), 15 °C/min. to 180 °C, 7 °C/min. to 230 °C, 30 °C/min. to 380 °C (10 min.) |
| Det.: | FID, 380 °C |
| Carrier Gas: | helium, 3.0 mL/min. |
| Injection: | 1 µL, cold on-column |
| Sample: | B100 Biodiesel plus Butanetriol Internal Standard (44896-U) and Tricaprin Internal Standard (44897-U), derivatized with MSTFA (394866) then diluted in heptane |