Merck
  • 1,3-dinitrobenzene induces age- and region-specific oxidation to mitochondria-related proteins in brain.
1,3-dinitrobenzene induces age- and region-specific oxidation to mitochondria-related proteins in brain.
Toxicological sciences : an official journal of the Society of Toxicology (2015-02-27)
Laura L Kubik, Rory W Landis, Henriette Remmer, Ingrid L Bergin, Martin A Philbert
ABSTRACT
Regions of the brain with high energy requirements are especially sensitive to perturbations in mitochondrial function. Hence, neurotoxicant exposures that target mitochondria in regions of high energy demand have the potential to accelerate mitochondrial damage inherently occurring during the aging process. 1,3-Dinitrobenzene (DNB) is a model neurotoxicant that selectively targets mitochondria in brainstem nuclei innervated by the eighth cranial nerve. This study investigates the role of age in the regional susceptibility of brain mitochondria-related proteins (MRPs) to oxidation following exposure to DNB. Male F344 rats (1 month old [young], 3 months old [adult], 18 months old [aged]) were exposed to 10 mg/kg DNB prior to mitochondrial isolation and histopathology experiments. Using a high-throughput proteomic approach, 3 important region- and age-related increases in DNB-induced MRP oxidation were determined: (1) brainstem mitochondria are ×3 more sensitive to DNB-induced oxidation than cortical mitochondria; (2) oxidation of brainstem MRPs is significantly higher than in cortical counterparts; and (3) MRPs from the brainstems of older rats are significantly more oxidized than those from young or adult rats. Furthermore, lower levels of DNB cause signs of intoxication (ataxia, chromodacryorrhea) and vacuolation of the susceptible neuropil in aged animals, while neither is observed in DNB-exposed young rats. Additionally, methemoglobin levels increase significantly in DNB-exposed adult and aged animals, but not young DNB-exposed animals. This suggests that oxidation of key MRPs observed in brainstem of aged animals is necessary for DNB-induced signs of intoxication and lesion formation. These results provide compelling evidence that environmental chemicals such as DNB may aid in the acceleration of injury to specific brain regions by inducing oxidation of sensitive mitochondrial proteins.
Product Description
Supelco
DL-Dithiothreitol solution, 1 M in H2O
SAFC
BIS-TRIS
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) trifluoroacetic acid, suitable for HPLC
Sigma-Aldrich
DL-Dithiothreitol solution, BioUltra, Molecular Biology, ~1 M in H2O
Sigma-Aldrich
BIS-TRIS, ≥98.0% (titration)
Sigma-Aldrich
BIS-TRIS, BioPerformance Certified, suitable for cell culture, suitable for insect cell culture, ≥98.0%
Sigma-Aldrich
BIS-TRIS, BioUltra, ≥99.0% (NT)
Sigma-Aldrich
BIS-TRIS, BioXtra, ≥98.0% (titration)
SAFC
BIS-TRIS
Sigma-Aldrich
Ultrapure Acetonitrile
SAFC
Iodoacetamide
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Iodoacetamide, BioUltra
Sigma-Aldrich
Iodoacetamide, ≥99% (NMR), crystalline
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Iodoacetamide, Single use vial of 56 mg
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
Dimethyl sulfoxide, Molecular Biology
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.6%, ReagentPlus®
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%