Merck
  • Yellow Pigment Aurovertins Mediate Interactions between the Pathogenic Fungus Pochonia chlamydosporia and Its Nematode Host.
Yellow Pigment Aurovertins Mediate Interactions between the Pathogenic Fungus Pochonia chlamydosporia and Its Nematode Host.
Journal of agricultural and food chemistry (2015-07-08)
Yan-li Wang, Lin-fang Li, Dong-xian Li, Baile Wang, Keqin Zhang, Xuemei Niu
ABSTRACT
Nematophagous fungi are globally distributed soil fungi and well-known natural predators of soil-dwelling nematodes. Pochonia chlamydosporia can be found in diverse nematode-suppressive soils as a parasite of nematode eggs and is one of the most studied potential biological control agents of nematodes. However, little is known about the functions of small molecules in the process of infection of nematodes by this parasitic fungus or about small-molecule-mediated interactions between the pathogenic fungus and its host. Our recent study demonstrated that a P. chlamydosporia strain isolated from root knots of tobacco infected by the root-knot nematode Meloidogyne incognita produced a class of yellow pigment metabolite aurovertins, which induced the death of the free-living nematode Panagrellus redivevus. Here we report that nematicidal P. chlamydosporia strains obtained from the nematode worms tended to yield a total yellow pigment aurovertin production exceeding the inhibitory concentration shown in nematicidal bioassays. Aurovertin D was abundant in the pigment metabolites of P. chlamydosporia strains. Aurovertin D showed strong toxicity toward the root-knot nematode M. incognita and exerted profound and detrimental effects on the viability of Caenorhabditis elegans even at a subinhibitory concentration. Evaluation of the nematode mutation in the β subunit of F1-ATPase, together with the application of RNA interference in screening each subunit of F1FO-ATPase in the nematode worms, demonstrated that the β subunit of F1-ATPase might not be the specific target for aurovertins in nematodes. The resistance of C. elegans daf-2(e1370) and the hypersensitivity of C. elegans daf-16(mu86) to aurovertin D indicated that DAF-16/FOXO transcription factor in nematodes was triggered in response to the aurovertin attack. These findings advance our understanding of the roles of aurovertin production in the interactions between nematodes and the pathogen fungus P. chlamydosporia.
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) trifluoroacetic acid, suitable for HPLC
Sigma-Aldrich
Formic acid, ≥95%, FCC, FG
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.6%, ReagentPlus®
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, Molecular Biology
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
5-Hydroxy-1,4-naphthoquinone, 97%
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Oligomycin A, ≥99% (HPLC)
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis