Merck
  • Personality Traits Are Associated With Cortical Development Across Adolescence: A Longitudinal Structural MRI Study.
Personality Traits Are Associated With Cortical Development Across Adolescence: A Longitudinal Structural MRI Study.
Child development (2018-01-10)
Lia Ferschmann, Anders M Fjell, Margarete E Vollrath, Håkon Grydeland, Kristine B Walhovd, Christian K Tamnes
ZUSAMMENFASSUNG
How personality traits relate to structural brain changes in development is an important but understudied question. In this study, cortical thickness (CT) and surface area (SA), estimated using magnetic resonance imaging (MRI), were investigated in 99 participants aged 8-19 years. Follow-up MRI data were collected after on average 2.6 years for 74 individuals. The Big Five personality traits were related to longitudinal regional CT or SA development, but limited cross-sectional relations were observed. Conscientiousness, emotional stability, and imagination were associated with more age-expected cortical thinning over time. The results suggest that the substantial individual variability observed in personality traits may partly be explained by cortical maturation across adolescence, implying a developmental origin for personality-brain relations observed in adults.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Thiophene-2, ≥98% (HPLC)