Merck
  • Responsive Self-Assembly of Supramolecular Hydrogel Based on Zwitterionic Liquid Asymmetric Gemini Guest.
Responsive Self-Assembly of Supramolecular Hydrogel Based on Zwitterionic Liquid Asymmetric Gemini Guest.
Chemistry (Weinheim an der Bergstrasse, Germany) (2018-04-24)
Aoli Wu, Panpan Sun, Na Sun, Liqiang Zheng
ZUSAMMENFASSUNG
A low-molecular-weight supramolecular hydrogel has been fabricated based on host-guest interactions between β-cyclodextrin (β-CD) and an asymmetric gemini zwitterionic liquid (ZIL) containing an azobenzene and bis(trifluoromethanesulfonyl)imide. Reversible sol-gel phase transitions were triggered by light and temperature. The binding stoichiometry, mainly noncovalent interactions in the hydrogel, photo- and thermoresponsive mechanisms, and mode of inclusion in the complex were studied in detail by NMR spectroscopy, UV/Vis spectroscopy, isothermal titration calorimetry, and control experiments. Interestingly, the β-CD in the complex can be photo-manipulated to shuttle along the guest molecule, accompanied by a change in the circular dichroism signals. Reversible switching of the conductivity accompanies the sol-gel phase transition, which demonstrates the stability of the supramolecular hydrogel and its potential application in multi-stimuli-responsive electric sensor materials.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
N-Bromsuccinimid, ReagentPlus®, 99%
Sigma-Aldrich
p-Toluidin, 99%
Sigma-Aldrich
1,3-Propansulton, ≥99%
Sigma-Aldrich
1-Bromdecan, 98%
Sigma-Aldrich
Nitrosobenzol, ≥97%