Merck
Inhibition of mitochondrial fission prevents cell cycle progression in lung cancer.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology (2012-02-11)
Jalees Rehman, Hannah J Zhang, Peter T Toth, Yanmin Zhang, Glenn Marsboom, Zhigang Hong, Ravi Salgia, Aliya N Husain, Christian Wietholt, Stephen L Archer
ZUSAMMENFASSUNG
Mitochondria exist in dynamic networks that undergo fusion and fission. Mitochondrial fusion and fission are mediated by several GTPases in the outer mitochondrial membrane, notably mitofusin-2 (Mfn-2), which promotes fusion, and dynamin-related protein (Drp-1), which promotes fission. We report that human lung cancer cell lines exhibit an imbalance of Drp-1/Mfn-2 expression, which promotes a state of mitochondrial fission. Lung tumor tissue samples from patients demonstrated a similar increase in Drp-1 and decrease in Mfn-2 when compared to adjacent healthy lung. Complementary approaches to restore mitochondrial network formation in lung cancer cells by overexpression of Mfn-2, Drp-1 inhibition, or Drp-1 knockdown resulted in a marked reduction of cancer cell proliferation and an increase in spontaneous apoptosis. The number of cancer cells in S phase decreased from 32.4 ± 0.6 to 6.4 ± 0.3% with Drp-1 inhibition (P<0.001). In a xenotransplantation model, Mfn-2 gene therapy or Drp-1 inhibition could regress tumor growth. The tumor volume decreased from 205.6 ± 59 to 70.6 ± 15 mm(3) (P<0.05) with Mfn-2 overexpression and from 186.0 ± 19 to 87.0 ± 6 mm(3) (P<0.01) with therapeutic Drp-1 inhibition. Impaired fusion and enhanced fission contribute fundamentally to the proliferation/apoptosis imbalance in cancer and constitute promising novel therapeutic targets.