Merck
  • A nuclear export signal in the N-terminal regulatory domain of IkappaBalpha controls cytoplasmic localization of inactive NF-kappaB/IkappaBalpha complexes.
A nuclear export signal in the N-terminal regulatory domain of IkappaBalpha controls cytoplasmic localization of inactive NF-kappaB/IkappaBalpha complexes.
Proceedings of the National Academy of Sciences of the United States of America (2000-02-03)
T T Huang, N Kudo, M Yoshida, S Miyamoto
ZUSAMMENFASSUNG
Appropriate subcellular localization is crucial for regulation of NF-kappaB function. Herein, we show that latent NF-kappaB complexes can enter and exit the nucleus in preinduction states. The nuclear export inhibitor leptomycin B (LMB) sequestered NF-kappaB/IkappaBalpha complexes in the nucleus. Using deletion and site-directed mutagenesis, we identified a previously uncharacterized nuclear export sequence in residues 45-54 of IkappaBalpha that was required for cytoplasmic localization of inactive complexes. This nuclear export sequence also caused nuclear exclusion of heterologous proteins in a LMB-sensitive manner. Importantly, a LMB-insensitive CRM1 mutant (Crm1-K1) abolished LMB-induced nuclear accumulation of the inactive complexes. Moreover, a cell-permeable p50 NF-kappaB nuclear localization signal peptide also blocked these LMB effects. These results suggest that NF-kappaB/IkappaBalpha complexes shuttle between the cytoplasm and nucleus by a nuclear localization signal-dependent nuclear import and a CRM1-dependent nuclear export. The LMB-induced nuclear complexes could not bind DNA and were inaccessible to signaling events, because LMB inhibited NF-kappaB activation without affecting the subcellular localization of upstream kinases IKKbeta and NIK. Our findings indicate that the dominant nuclear export over nuclear import contributes to the largely cytoplasmic localization of the inactive complexes to achieve efficient NF-kappaB activation by extracellular signals.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Leptomycin B -Lösung aus Streptomyces sp., ≥95% (HPLC), Supplied in methanol: water (7:3)