Merck
  • Effect of the scaffold microenvironment on cell polarizability and capacitance determined by probabilistic computations
Effect of the scaffold microenvironment on cell polarizability and capacitance determined by probabilistic computations
Bio-medical materials and engineering (2018)
Betancourt BA, et al.