Merck
Dissipation and Risk Assessment of Multiresidual Fungicides in Grapes under Field Conditions.
Journal of agricultural and food chemistry (2019-12-17)
Mei Yang, Fengjian Luo, Xinzhong Zhang, Li Zhou, Zhengyun Lou, Meiqin Zhao, Zongmao Chen
ZUSAMMENFASSUNG
Grapes are among the most popular fruits globally, and various fungicides are widely applied to grape crops. As such, the presence of multiple fungicide residues and dietary risks in grapes has become the focus of significant attention. In this study, an easy-to-implement and sensitive UPLC-MS/MS approach was developed to simultaneously determine pyraclostrobin, dimethomorph, cymoxanil, cyazofamid and its metabolite CCIM in grapes via QuEChERS. This approach achieved 78.1-106.0% recovery and a 0.01 mg kg-1 limit of quantitation (LOQ). Field trials revealed that these compounds had degradation half-lives ranging from 0.9 to 13.3 days. And their terminal residues ranging from < LOQ to 1.36 mg kg-1 were below the official maximum residue limit (MRL) in China. The short-term risk for each tested fungicide was below 54%. The long-term risk of individual chemicals ranged from 0.0086% to 3.1%, and their cumulative risk was 4.4%. Results indicated that the dietary risk of these fungicides in grapes was minor.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Supelco
Cymoxanil, PESTANAL®, analytical standard