Merck
Role of opioid peptides in the regulation of cytokine production by murine CD4+ T cells.
Cellular immunology (1994-03-01)
P van den Bergh, R Dobber, S Ramlal, J Rozing, L Nagelkerken
ZUSAMMENFASSUNG
The presence of the opioid peptides alpha- and beta-endorphin (-End) but not methionine enkephalin (Met-enk) in in vitro cultures of purified CD4+ T cells, stimulated with concanavalin A in the presence of irradiated spleen cells, resulted in a threefold stimulation of IL-2, IL-4, and IFN-gamma production. The stimulating effect was dependent on the concentration of the peptides and reached optimal values in the dose range from 10(-12) to 10(-10) M. Similar results were obtained when purified CD4+ T cells were stimulated with immobilized anti-CD3, indicating a direct effect of opioid peptides on CD4+ T cells. Moreover, in this system a twofold enhancement of IL-6, but not IL-1, secretion was observed. These stimulatory effects were not mediated through opioid receptors since the peptide fragment beta-End6-31 that lacks the N-terminal opioid receptor binding part was still stimulatory. This is in agreement with our finding that beta-End did not affect cAMP, as described for the triggering of classical opioid receptors. Experiments undertaken to reveal the mechanism of action of opioid peptides suggest an overall enhancement of lymphokine production: (1) enhancement of IL-4 production occurred also in the presence of excess IL-2; and (2) neither IL-1 receptor-antagonizing protein nor anti-IL-6 were capable to abrogate the stimulatory effect on IL-2 and IL-4 production. Finally, the presence and activity of opioid receptors in cultures of CD4+ T cells were substantiated by the fact that the opioid receptor antagonist naloxone by itself enhanced cytokine synthesis, which points to the endogenous production by lymphocytes of down-regulating opioid peptides.