Merck
CaMKII regulates diacylglycerol lipase-α and striatal endocannabinoid signaling.
Nature neuroscience (2013-03-19)
Brian C Shonesy, Xiaohan Wang, Kristie L Rose, Teniel S Ramikie, Victoria S Cavener, Tyler Rentz, Anthony J Baucum, Nidhi Jalan-Sakrikar, Ken Mackie, Danny G Winder, Sachin Patel, Roger J Colbran
ZUSAMMENFASSUNG
The endocannabinoid 2-arachidonoylglycerol (2-AG) mediates activity-dependent depression of excitatory neurotransmission at central synapses, but the molecular regulation of 2-AG synthesis is not well understood. Here we identify a functional interaction between the 2-AG synthetic enzyme diacylglycerol lipase-α (DGLα) and calcium/calmodulin dependent protein kinase II (CaMKII). Activated CaMKII interacted with the C-terminal domain of DGLα, phosphorylated two serine residues and inhibited DGLα activity. Consistent with an inhibitory role for CaMKII in 2-AG synthesis, in vivo genetic inhibition of CaMKII increased striatal DGL activity and basal levels of 2-AG, and CaMKII inhibition augmented short-term retrograde endocannabinoid signaling at striatal glutamatergic synapses. Lastly, blockade of 2-AG breakdown using concentrations of JZL-184 that have no effect in wild-type mice produced a hypolocomotor response in mice with reduced CaMKII activity. These findings provide mechanistic insights into the molecular regulation of striatal endocannabinoid signaling with implications for physiological control of motor function.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Lipoprotein-Lipase aus Kuhmilch, ammonium sulfate suspension, ≥2,000 units/mg protein (BCA)
Sigma-Aldrich
Lipoprotein-Lipase aus Pseudomonas sp., lyophilized, powder, ≥1200 U/mg
Sigma-Aldrich
Lipoprotein-Lipase aus Pseudomonas sp., lyophilized powder, ≥50,000 units/mg solid