Merck
  • Interaction of human DNA polymerase eta with monoubiquitinated PCNA: a possible mechanism for the polymerase switch in response to DNA damage.
Interaction of human DNA polymerase eta with monoubiquitinated PCNA: a possible mechanism for the polymerase switch in response to DNA damage.
Molecular cell (2004-05-20)
Patricia L Kannouche, Jonathan Wing, Alan R Lehmann
ZUSAMMENFASSUNG
Most types of DNA damage block replication fork progression during DNA synthesis because replicative DNA polymerases are unable to accommodate altered DNA bases in their active sites. To overcome this block, eukaryotic cells employ specialized translesion synthesis (TLS) polymerases, which can insert nucleotides opposite damaged bases. In particular, TLS by DNA polymerase eta (poleta) is the major pathway for bypassing UV photoproducts. How the cell switches from replicative to TLS polymerase at the site of blocked forks is unknown. We show that, in human cells, PCNA becomes monoubiquitinated following UV irradiation of the cells and that this is dependent on the hRad18 protein. Monoubiquitinated PCNA but not unmodified PCNA specifically interacts with poleta, and we have identified two motifs in poleta that are involved in this interaction. Our findings provide an attractive mechanism by which monoubiquitination of PCNA might mediate the polymerase switch.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Taq-DNA-Polymerase aus Thermus aquaticus, with 10× PCR reaction buffer containing MgCl2
Sigma-Aldrich
Taq-DNA-Polymerase aus Thermus aquaticus, with 10× PCR reaction buffer without MgCl2
Sigma-Aldrich
DNA-Polymerase I aus E. coli lysogen für NM 964, buffered aqueous glycerol solution