Merck
  • Synthesis of D-mannitol and L-iditol derivatives as monomers for the preparation of new regioregular AABB-type polyamides.
Synthesis of D-mannitol and L-iditol derivatives as monomers for the preparation of new regioregular AABB-type polyamides.
Carbohydrate research (2002-03-23)
Manuel Mancera, Isaac Roffé, Manuel Rivas, Carmen Silva, Juan A Galbis
ZUSAMMENFASSUNG
1,6-Diamino-1,6-dideoxy-2,3,4,5-tetra-O-methyl-D-mannitol (and its L-iditol analogue) suitable for their utilization as monomers in the preparation of linear polyamides are described. Regio- and stereoregular polyamides of the AABB-type have been prepared by the active ester polycondensation method from these C(2) symmetric monomers and suberic and dodecanedioic acids. The resulting polyamides were obtained in fair yields (70-60%) and were characterized by elemental analyses and infrared and 1H and 13C NMR spectroscopies. Their M(w) and M(w)/M(n) were determined by GPC relative to polystyrene standards. All of them were gummy non-crystalline solids.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Dodecandisäure, 99%
Sigma-Aldrich
Korksäure, 98%
Sigma-Aldrich
Korksäure, purum, ≥98.0% (T)
Sigma-Aldrich
L-Iditol, ≥98% (GC)