Merck
[Studies on the chemical constituents of extract with water from Forsythia suspensa].
Zhong yao cai = Zhongyaocai = Journal of Chinese medicinal materials (2010-06-26)
Lan Luan, Gang-li Wang, Rui-chao Lin
ZUSAMMENFASSUNG
To study the chemical constituents of the extract with water from Forsythia suspensa. The compounds were isolated and repeatedly purified on TLC, silica gel column chromatograph, gel column chromatography, and preparative HPLC, and the structures were elucidated by physico-chemical properties and NMR. Eight compounds were obtained and elucidated as stearic acid(I), ursolic acid(II),p-hydroxyphenyl acetic acid(III),3-(4-ehtoxoy-3-hydroxyphenyl)acrylic acid(IV),isolariciresinol(V), epipinoresinol-4-O-beta-D-glucoside(VI),(+)-epipinoresionl(VII),(+)-pinoresinol(VIII). Compound III is isolated from this genus for the first time.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
4-Hydroxy-phenylessigsäure, 98%