Merck
Anethole isomerization and dimerization induced by acid sites or UV irradiation.
Molecules (Basel, Switzerland) (2010-07-27)
Hans T Castro, Jairo René Martínez, Elena Stashenko
ZUSAMMENFASSUNG
The formation of cis-anethole and various dimers as a result of the exposure of trans-anethole to microporous solid acids (dealuminated HY zeolites), or UV-Vis irradiation was established by means of high resolution gas chromatography coupled to mass spectrometry. 3,4-bis-(4-Methoxyphenyl)-(E)-hex-2-ene was the most abundant compound among eight different methoxyphenyl-disubstituted hexenes produced by electrophilic addition and elimination reactions induced by HY zeolites. (1a,2a,3b,4b)-1,2-bis(4-Methoxyphenyl)-3,4-dimethylcyclobutane was the principal component in the mixture of 5 methoxyphenyl-disubstituted cyclobutanes found, together with cis-anethole, after UV-Vis irradiation of a trans-anethole solution in toluene.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
trans-Anethol, 99%
Sigma-Aldrich
trans-Anethol, ≥99%, FCC, FG
Supelco
trans-Anethol, analytical standard
Supelco
trans-Anethol, Pharmaceutical Secondary Standard; Certified Reference Material