Merck
  • Preparation, spectroscopy, EXAFS, electrochemistry and pharmacology of new ruthenium(II) carbonyl complexes containing ferrocenylthiosemicarbazone and triphenylphosphine/arsine.
Preparation, spectroscopy, EXAFS, electrochemistry and pharmacology of new ruthenium(II) carbonyl complexes containing ferrocenylthiosemicarbazone and triphenylphosphine/arsine.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy (2011-01-11)
R Prabhakaran, S Anantharaman, M Thilagavathi, M V Kaveri, P Kalaivani, R Karvembu, N Dharmaraj, H Bertagnolli, F Dallemer, K Natarajan
ZUSAMMENFASSUNG
A new series of new hetero-bimetallic complexes containing iron and ruthenium of the general formula [RuCl(CO)(B)(EPh3)(L)] (where E=P or As; B=PPh3, AsPh3, py or pip; L=ferrocene derived monobasic bidentate thiosemicarbazone ligand) have been synthesized by the reaction between ferrocene-derived thiosemicarbazones and ruthenium(II) complexes of the type [RuHCl(CO)(B)(EPh3)2] (where E=P or As; B=PPh3, AsPh3, py or pip). The new complexes have been characterized by elemental analyses, IR, electronic, NMR (1H, 13C and 31P), EXAFS (extended X-ray absorption fine structure spectroscopy) and cyclic voltammetric techniques. Antibacterial activity of the new complexes has been screened against Escherichia coli, Vibrio cholerae, and Pseudomonas aeruginosa species.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Triphenylphosphin, ReagentPlus®, 99%
Sigma-Aldrich
Triphenylphosphin, ≥95.0% (GC)