Merck
  • Ammonia as a preferred additive in chiral and achiral applications of supercritical fluid chromatography for small, drug-like molecules.
Ammonia as a preferred additive in chiral and achiral applications of supercritical fluid chromatography for small, drug-like molecules.
Journal of chromatography. A (2011-12-22)
Manuel Ventura, Brent Murphy, Wolfgang Goetzinger
ZUSAMMENFASSUNG
Supercritical fluid chromatography is routinely utilized by analytical separations groups in the pharmaceutical industry to efficiently handle separations for discovery medicinal chemistry purposes. Purifications are performed on samples ranging from a few milligrams up to hundreds of grams. Basic additives dissolved into the liquid component of the SFC mobile phase are commonly used to improve peak shape and efficiency in achiral and chiral separations. While for purposes of analysis there is minimal consequence to additive introduction in the mobile phase, for preparative separations one needs to consider the potential effect of an additive's presence when concentrated with the desired compound. Following an SFC purification using an additive-containing modifier, the resulting fractions will contain an easily evaporated modifier, and after its evaporation perhaps still significant levels of the less volatile additive. Depending on the aqueous solubility and basicity of the final product, the process of removing basic amine additives can be time-consuming and can result in reduced yields. NMR analysis following preparative isolation and evaporation often reveals the fact of insufficient removal of the chromatographic additive even after aqueous work up steps. In this study, ammonia is evaluated as an alternative additive to strong bases such as diethylamine (DEA) in SFC purification and analysis and to the authors' knowledge no previous publication has been written describing the application of methanolic ammonia as an additive for SFC separations. Dimethylethylamine (DMEA), a more volatile additive than DEA, is also evaluated relative to ammonia for its potential to simplify the isolation process after purification and in terms of chromatographic performance. The loss in concentration of ammonia in methanol modifier over time due to evaporation and effects of that loss are also described. Furthermore, for ammonia the analytical benefit is shown to extend to on-line mass spectrometric detection relative to other basic additives.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Diethylamin, ≥99.5%
Sigma-Aldrich
Diethylamin, puriss. p.a., ≥99.5% (GC)
Sigma-Aldrich
Diethylamin, purified by redistillation, 99.5%
Sigma-Aldrich
N,N-Dimethylethylamin, ≥99%
Sigma-Aldrich
Diethylamin -hydrochlorid, ReagentPlus®, 99%
Sigma-Aldrich
2-Ethylpyridin, 97%
Sigma-Aldrich
N,N-Dimethylethylamin, 99%
Sigma-Aldrich
Diethylamin -hydrobromid, 98%