Merck
  • Lead exposure in pheochromocytoma cells induces persistent changes in amyloid precursor protein gene methylation patterns.
Lead exposure in pheochromocytoma cells induces persistent changes in amyloid precursor protein gene methylation patterns.
Environmental toxicology (2012-07-06)
Yuan-Yuan Li, Tian Chen, Yanjian Wan, Shun-qing Xu
ZUSAMMENFASSUNG
It has been suggested that lead (Pb) exposure in early life may increase amyloid precursor protein (APP) expression and promote the pathogenesis of Alzheimer's disease in old age. The current study examined whether the DNA methylation patterns of APP gene in rat pheochromocytoma (PC12) cells changed after Pb acetate exposure. Undifferentiated PC12 cells were exposed to three doses of Pb acetate (50, 250, and 500 nM) and one control for 2 days or 1 week. The methylation patterns of APP promoter and global DNA methylation were analyzed. The DNA methyltransferase 1 (DNMT1) expression and the level of amyloid β peptide (Aβ) were also investigated. The results showed that the exposure of the three concentrations of Pb acetate could make the APP promoter hypomethylated. The global DNA methylation level and the expression of DNMT1 were changed in the 500 nM group after 2 days exposure and in the 250 and 500 nM group after 7 days exposure. Thus, Pb may exert neurotoxic effects through mechanisms that alter the global and promoter methylation patterns of APP gene. © 2010 Wiley Periodicals, Inc. Environ Toxicol, 2012.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Blei(II)-acetat Trihydrat, ACS reagent, ≥99%
Sigma-Aldrich
Blei(II)-acetat Trihydrat, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., 99.5-102.0%
Sigma-Aldrich
Blei(II)-acetat Trihydrat, 99.999% trace metals basis
Sigma-Aldrich
Blei(II)-acetat Trihydrat, ≥99.99% trace metals basis