Merck
  • Methylopila musalis sp. nov., an aerobic, facultatively methylotrophic bacterium isolated from banana fruit.
Methylopila musalis sp. nov., an aerobic, facultatively methylotrophic bacterium isolated from banana fruit.
International journal of systematic and evolutionary microbiology (2012-09-18)
Nina V Doronina, Elena N Kaparullina, Tatjana V Bykova, Yuri A Trotsenko
ZUSAMMENFASSUNG
A newly isolated, facultatively methylotrophic bacterium (strain MUSA(T)) was investigated. The isolate was strictly aerobic, Gram-stain-negative, asporogenous, motile, rod-shaped and multiplied by binary fission. The strain utilized methanol, methylamine and an apparently narrow range of multi-carbon compounds, but not methane, dichloromethane or CO2/H2, as the carbon and energy sources. Growth occurred at pH 5.5-9.5 (optimum, pH 7.0) and 16-40 °C (optimum, 28-30 °C). The major fatty acids of methanol-grown cells were C18 : 1ω7c, C18 : 0 and 11-methyl-C18 : 1ω7c . The predominant phospholipids were phosphatidylethanolamine, phosphatidylcholine, phosphatidylglycerol and phosphatidylmonomethylethanolamine. The major ubiquinone was Q-10. The strain had methanol and methylamine dehydrogenases as well as the enzymes of the N-methylglutamate pathway (lyases of γ-glutamylmethylamide and N-methylglutamate). C1 assimilation occurs via the isocitrate lyase-negative serine pathway. Ammonium was assimilated by glutamate dehydrogenase and the glutamate cycle (glutamate synthase/glutamine synthetase). The DNA G+C content of the strain was 64.5 mol% (determined from the melting temperature). Based on 16S rRNA gene sequence similarity (97.0-98.9 %) and DNA-DNA relatedness (36-38 %) with representatives of the genus Methylopila (Methylopila capsulata IM1(T) and Methylopila jiangsuensis JZL-4(T)) the isolate was classified as a novel species of the genus Methylopila, for which the name Methylopila musalis sp. nov. is proposed. The type strain is MUSA(T) ( = VKM B-2646(T) = DSM 24986(T) = CCUG 61696(T)).
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Coenzyme Q2, ≥90%