Merck
TLR4 is a target of environmentally relevant concentration of lead.
Toxicology letters (2012-09-25)
Ana L Luna, Leonor C Acosta-Saavedra, Marcela Martínez, Nallely Torres-Avilés, Rocío Gómez, Emma S Calderón-Aranda
ZUSAMMENFASSUNG
Lead (Pb) alters the susceptibility to different pathogens suggesting that macrophage-mediated defense mechanisms, through activation of toll-like receptors (TLRs), may be affected by Pb. The aim of this study was to test whether activation of TLR4 is a targeted molecule to the effect of environmentally relevant Pb concentrations (0.05, 0.5 and 5μg/dL). The function of macrophages activated through TLR4 was evaluated using as TLR4 ligand lipopolysaccharides (LPSs) from two different pathogens: Escherichia coli and Salmonella typhimurium. Pb induced proliferation, increased the NO(-) baseline, IL-1β and IL-6 secretion. Interestingly, Pb exposure induced differential effects on cells stimulated with the two LPS used: in macrophages stimulated with LPS from E. coli, Pb caused an early decrease in proliferation, increase NO(-) production, and decrease IL-6 and TNF-α secretion; in macrophages stimulated with LPS from S. typhimurium, Pb decreased proliferation after 36h, induced a biphasic effect on NO(-) production, and enhance the secretion of IL-1β, IL-6 and TNF-α. Results suggest that TLR4 is a target for the Pb effect, which up to 5.0μg/dL affect immune competence against pathogens, dependent on the bacterial species. This effect may be attributable to structural differences that determine LPS affinity for TLR4.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Blei(II)-acetat Trihydrat, ACS reagent, ≥99%
Sigma-Aldrich
Blei(II)-acetat Trihydrat, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., 99.5-102.0%
Sigma-Aldrich
Blei(II)-acetat Trihydrat, 99.999% trace metals basis
Sigma-Aldrich
Blei(II)-acetat Trihydrat, ≥99.99% trace metals basis