Merck
  • [Preparation and applications of a supported liquid-liquid extraction column with a composite diatomite material].
[Preparation and applications of a supported liquid-liquid extraction column with a composite diatomite material].
Se pu = Chinese journal of chromatography (2012-12-22)
Jianmin Bao, Zhishuang Ma, Ying Sun, Yongzun Wang, Youxin Li
ZUSAMMENFASSUNG
A rapid and special supported liquid-liquid extraction (SLE) column was developed with a composite diatomite material. The SLE column was evaluated by high performance liquid chromatography (HPLC) with acidic, neutral and alkaline compounds dissolved in water. Furthermore, some real complex samples were also analyzed by HPLC with the SLE method. The recoveries of benzoic acid (acidic), p-nitroaniline (alkaline) and 4-hydroxy-benzoic methyl ester (neutral) treated by the SLE column were 90.6%, 98.1% and 97.7%. However, the recoveries of the three compounds treated by traditional liquid-liquid extraction (LLE) method were 71.9%, 81.9% and 83.9%. The results showed that the SLE technique had higher recoveries than the traditional LLE method. The spiked recoveries of the complex samples, such as benzoic acid in Sprite and dexamethasone acetate, chlorphenamine maleate, indomethacin in bovine serum, were between 80% and 110% and the relative standard deviations (RSDs) were less than 15%. For biological specimen, the results could be accepted. Meantime, many disadvantages associated with traditional LLE method, such as emulsion formation, didn't occur using SLE column. The SLE column technique is a good sample preparation method with many advantages, such as rapid, simple, robust, easily automated, high recovery and high-throughput, which would be widely used in the future.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Celite® 545, filter aid, treated with sodium carbonate, flux calcined
Sigma-Aldrich
Benzoesäure, ACS reagent, ≥99.5%
Sigma-Aldrich
Filtriermittel, Celite® 545
Supelco
Benzoesäure, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Celatom®, säuregewaschen, for use in Total Dietary Fiber Assay, TDF-100A
Sigma-Aldrich
Celite® S, filter aid, dried, untreated
Sigma-Aldrich
Hyflo® Super Cel®, filter aid, flux calcined, treated with sodium carbonate
Supelco
Mettler-Toledo Kalibriersubstanz ME 18555, Benzoesäure, analytical standard, (for the calibration of the melting point system), traceable to primary standards (LGC)
Supelco
Schmelzpunktstandard 121-123°C, analytical standard
Sigma-Aldrich
Diatomeenerde
Sigma-Aldrich
Benzoesäure, meets analytical specification of Ph. Eur., BP, USP, FCC, E210, 99.5-100.5% (alkalimetric)
Sigma-Aldrich
Celite® 545 AW, filter aid, acid washed, treated with sodium carbonate, flux calcined
Supelco
Celite® 545 AW, reagent grade
Sigma-Aldrich
Kieselgur, ungewaschen, powder
Supelco
Benzoesäure, reference material for titrimetry, certified by BAM, >99.5%
Sigma-Aldrich
Celpure® P1000, meets USP/NF testing specifications
Sigma-Aldrich
Celite® 535, filter aid, flux calcined
Sigma-Aldrich
Benzoesäure, ≥99.5%, FCC, FG
Sigma-Aldrich
Celite® 503, filter aid, treated with sodium carbonate, calcined
Sigma-Aldrich
Benzoesäure, ReagentPlus®, 99%
Sigma-Aldrich
Benzoesäure, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99.9% (alkalimetric)
Supelco
Benzoesäure, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Dexamethason-21-Acetat, ≥99%
Sigma-Aldrich
Benzoesäure, natural, ≥99.5%, FCC, FG
Sigma-Aldrich
Kaliumbenzoat, ReagentPlus®, 99%
Sigma-Aldrich
Benzoesäure, purified by sublimation, ≥99%
Dexamethasonacetat, European Pharmacopoeia (EP) Reference Standard